Organischer Referenzstandard, 2-Isopropyl-3-methoxypyrazin 100 µg/ml in Methanol, Supelco®
Catalog # 47527-U
Supplier: Merck
CAS Number:
Specifications
- VE:1
- Normen:Certified reference material
- CAS Nr.:25773-40-4
- Bezeichnung:2-Isopropyl-3-methoxypyrazin 100 µg/ml in Methanol
- Lagerbedingungen:2...30 °C
- Beschreibung Gefahrenklasse:H225 - H301 + H311 + H331 - H370
- Shipping conditions:STD
- M-Clarity Qualitätsstufe:MQ100
- Flammpunkt:11 °C
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311