Microtubes with screw cap
: BIOTIX
PP, with PE cap
- Leak proof seal to –80 °C
- DNase, RNase and pyrogen free
- Centrigugation 20000×g for 15 minutes
- Autoclavable
Ideal for centrifugation to PCR and freezer storage.
Frequently Bought Together
PP, with PE cap
Ideal for centrifugation to PCR and freezer storage.