Pipette tips, gel loading
Supplier: BIOTIX
Certificates
About this item
These pipette tips are specially designed to help load the thinnest sequencing gels.
- Extended length
- Universal fit: Ideal for single & multi-channel pipettors
- Eco-friendly racks
Frequently Bought Together