Digoxin
Glass bottle for solids, 1 mg
Catalog # FITZ10-D05-M91281
Supplier: FITZGERALD INDUSTRIES INTL
Jump to:
Product Details & Documents
- CAS Number:20830-75-5
- Formula:C₄₁H₆₄O₁₄
- MDL Number:MFCD00003674
- Molecular Weight:780.95 g/mol
- EINECS:244-068-1
- UN:2811
- ADR:6.1,II
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:1 mg