ContactUsLinkComponent
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂), Sigma-Aldrich®
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂), Sigma-Aldrich®
Catalog # SIALN6785-10VL
Supplier:  Merck
CAS Number:  
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂), Sigma-Aldrich®
Catalog # SIALN6785-10VL
Supplier:  Merck
CAS Number:  
Specifications
  • VE:
    10 Vial
  • Seq:
    0239
Specifications
About this item
 :
 :709.41 g/mol
EINECS:210-123-3
CAS Number:606-68-8
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂
MDL Number:MFCD00036200