Multimode columns, Asahipak GS-HQ series preparative type
: Resonac
Asahipak GS-HQ series are polyvinyl alcohol-based columns.
- SEC is the main separation mode
- Depends on the eluent selected, the column adds multimode features of reversed phase, HILIC and ion exchange modes to SEC
- Suitable for the separation of peptides or nucleic acids with similar molecular weights
- Suitable for desalting samples or substituting buffer in protein analysis
- Catalog No:
- SHOWF6810035
- SHOWF6810034
- Bezeichnung:
- Asahipak GS-320 20G
- Asahipak GS-220 20G
- pH-Bereich:
- 2 - 12
- 2 - 9
- Temperaturlimit:
- 4 - 60 °C
- 4 - 60 °C
Frequently Bought Together