AuftragsverfolgungKontakt
Enterotomy scissors
Enterotomy scissors
Geknöpft/stumpfe, 215 mm
Catalog # MART11-784-21
Supplier:   KLS Martin Group
Enterotomy scissors
Catalog # MART11-784-21
Supplier:  KLS Martin Group
Enterotomy scissors
Geknöpft/stumpfe, 215 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Enterotomy scissors made of stainless steel 18/8.
Specifications
  • Typ:
    Geknöpft/stumpfe
  • Länge:
    215 mm