Tube rack, for water bath, SP Bel-Art
BELAF188820000
- Für Röhrchen:1,5 − 15 ml
- Anz. Bohrungen:6
- Bezeichnung:Rack for water bath
- Material:PP
- VE:1
Height-adjustable racks securely hold up to six tubes or vials at any desired depth in a water bath or beaker. The rack has a flexible grip that easily retains mixed test tubes of varying weight and volume from 1,5 ml to 15 ml.
- Made of PP
- Adjustable height prevents submersion problems
- Compact footprint
- Stable base
The compact yet stable footprint saves bench and bath space and lifts in and out of a bath without splashing.