Microcentrifuge tube opener, SP Bel-Art
Catalog # BELAF199250001
Supplier: Bel-Art Products, a Part of SP
Specifications
- Bezeichnung:Microcentrifuge tube opener
- VE:3
Specifications
About this item
Easily open all sizes of microcentrifuge tubes with this double-ended tool.
- Opens standard 1,5, 2,0 and 2,2 ml tubes with one end
- Forked blade on other end opens smaller sizes (250, 400 and 550 μl)
- Dimensions: 7 cm (3⅛") long