Pestizid- Referenzstandard, 3,4,5-Trichlorphenol 1.000 µg/ml in Aceton
Catalog # ACCUM-1653-IS-R
Supplier: AccuStandard
CAS Number:
Specifications
- Packung:Glasflasche
- VE:1 mL
- CAS Nr.:609-19-8
- Bezeichnung:3,4,5-Trichlorphenol 1.000 µg/ml in Aceton
- Formel:C₆H₃Cl₃O
- Bezeichnung:3,4,5-Trichlorophenol