ContactUsLinkComponent
Holder for chromatography column, SP Bel-Art
Holder for chromatography column, SP Bel-Art
Catalog # BELAF182020000
undefined
Holder for chromatography column, SP Bel-Art
Catalog # BELAF182020000
Specifications
  • Bezeichnung:
    Holder, for eight 30 mm chromatography columns, 311×64 mm
  • VE:
    1
Specifications
About this item
Securely holds as many as eight chromatography columns up to 30 mm (1³/₁₆") in diameter in or between its flexible loops.
  • Rigid plastic construction
  • Easy to secure
  • Can stand alone or be wall-mounted
The rigid plastic back is mounted on two polyethylene support blocks which slide over support stand rods. Tighten the screws to mount the rack at the desired height. Requires two support rods and stands (H18303-0000, sold separately) or can be wall-mounted by removing the support blocks from the back of the unit.