Standard solution, phosphate, 500 mg/L as PO4 (NIST), pk/16 - 10-mL voluette® ampules
Catalog # 705-0058
Supplier: Hach
CAS Number:
Specifications
- VE:16
- Bezeichnung:Phosphate standard, 500 mg/l in 10 ml ampoules
- Pk Info:16 ampoules
Specifications
About this item
For accuracy checks in phosphate (phosphorus) determinations
- Simple Accuracy Checks
- Minimize waste
- Reliable results