Sampling spoon, large, Sterileware®, SP Bel-Art
Catalog # BELAH369480000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Kapazität:30 ml
- Länge:254 mm
- Material:PS
- VE:25
Specifications
About this item
PS, white. This large disposable spoon made from FDA-grade PS is ideal for sampling semi-solids, viscous liquids or bulk materials.
- Long handle reaches into containers and keeps hand away from sample
- Each one-piece spoon is sealed in a polyethylene bag, ready for use
- Sterilised by Gamma Irradiation
- Lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilisation