Biozid B, Premier Klercide-CR™
Artikelnr. 148-0070
Lieferant: ECOLAB CONTAMINATION CONTROL
Spezifikationen
- Kapazität:1 l
- Bezeichnung:Klercide™ Sporicidal Chlorine/Quat, sterile filtered biocide, spray
- Seq:01
- Verpackt:1 L bottle with trigger spray
- VE:6
Spezifikationen
Über diesen Artikel
Klercide™ Sporicidal Chlorine/Quat is a fast acting sporicide. Suitable for ISO Class 5; Grade A and B (microbial count) areas: cleanroom processed and sterile, double bagged. Blend of stabilised chlorine dioxide and quaternary ammonium compound, with broad spectrum activity. Filtered to 0,2 µm.
- Safe: non toxic and non corrosive, and very effective against bacterial spore
- CEN and isolate tested
- Fully compatible and ideal for a rotational regime
- SteriShield Delivery System™ ensures 6 months in-use shelf life
Certifications: EN standards: EN 1276 - bactericidal activity, EN 1650 - fungicidal activity, EN 13697 bactericidal surface test, EN 13704 sporocidal activity.
Caution: Use biocides safely. Always read the label and product information before use.