AuftragsverfolgungKontakt
Inbedmatrix OCT - Cryo | Avantor
Inbedmatrix OCT - Cryo
Cryotomy embedding matrix, Dispenser bottle, 125 mL
Artikelnr. 720-2356
Anbieter:   CELLPATH
Inbedmatrix OCT - Cryo
Artikelnr. 720-2356
Anbieter:  CELLPATH
CAS Nummer:  
Inbedmatrix OCT - Cryo
Inbedmatrix OCT - Cryo
Cryotomy embedding matrix, Dispenser bottle, 125 mL
CAS Nummer:  
Preis
22,80 €
Einloggen für Verfügbarkeit
1 * 125 mL
Product Details & Documents
‘OCT’ cryo microtomy embedding medium is designed for use at –20 ˚C to –30 ˚C. The medium freezes rapidly with minimal damage by crystal formation and is an ideal support medium for cutting tissue sections.
  • Liquid level clearly visible in bottle
  • Excellent quality, highly viscous
  • Easy to use dispenser bottle
‘OCT’ is easily removed from the microscope slide prior to staining. The 125 ml bottle is a convenient size with a nozzle moulded into the cap.
Spezifikationen
Spezifikationen
  • Packung:
    Dispenser bottle
  • VE:
    125 mL
  • Bezeichnung:
    Cryotomy embedding matrix
Spezifikationen