3-Propylphenol
Glass bottle for solids, 25 g
Artikelnr. MOLE28380943-25G
Marketsource
Anbieter: Molekula
Springe zu:
Product Details & Documents
Synonyme: 3-N-Propylphenol
- CAS Nummer:621-27-2
- Formel:C₉H₁₂O
- MDL-Nummer:MFCD01632130
- Molekulargewicht:136.19 g/mol
- EINECS:210-675-5
- UN:3145
- ADR:8,III
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:25 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



