Order Entry
Austria
ContactUsLinkComponent
4-Chloranilin, Supelco®
Toxic
Health
Environment
4-Chloranilin, Supelco®
Catalog # 35823-1G
Supplier:  Merck
CAS Number:  
undefined
Toxic
Health
Environment
4-Chloranilin, Supelco®
Catalog # 35823-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Merck Index:13,02136
Boiling Pt:232 °C (1013 hPa)
Melting Pt:69…71 °C
Density:1,43 g/cm³ (20 °C)
Flash Pt:120 °C
ADR:6.1,II
 :M-Clarity™ Qualitätsstufe = MQ100
 :127.57 g/mol
EINECS:203-401-0
UN:2018
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Raumtemperatur
CAS Number:106-47-8
Formula:C₆H₆ClN
MDL Number:MFCD00007835