Natriumdecylsulfat, Supelco®
Catalog # 71443-10G
Supplier: Merck
CAS Number:
Specifications
- Packung:Glasflasche für Feststoffe
- VE:10 g
- Seq:3236
Specifications
About this item
:
:260.33 g/mol
Melting Pt:191.85 °C
Density:1.207 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Raumtemperatur
EINECS:205-568-5
CAS Number:142-87-0
Formula:C₁₀H₂₁NaO₄S
MDL Number:MFCD00041881