(+)-Camphen
Glass bottle for solids, 500 g
Artikelnr. ROTH7558.1
Marketsource
Anbieter: Roth Carl
Springe zu:
Product Details & Documents
Synonyme: (1R)-(+)-2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane, (+)-3,3-Dimethyl-2-methylennorcamphane
- CAS Nummer:5794-03-6
- Formel:C₁₀H₁₆
- Molekulargewicht:136.24 g/mol
- Dichte:0,85 g/cm³ (20 °C)
- ADR:4.1,II
- :Density: 0.85 g/cm³ (20 °C)
- EINECS:227-336-2
- UN:1325
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:500 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



