Mikroplatten, Standardhöhe, 384 ShallowWell, Nunc™
Catalog # 734-2877
Supplier: Thermo Fisher Scientific
Specifications
- Anz. Wells:384
- Empf. Arbeitsvolumen:2 - 20 μl
- Steril:-
- Farbe:Schwarz
- Bezeichnung:384-ShallowWell-Platte, unbehandelte Oberfläche, ohne Deckel
- Environmentally Preferable:Umweltschonende Herstellung
- Umweltzertifizierung:NAR Certificate of Retirement
- Arbeitsvolumen:2-20 μl
- VE:100
- Wellvolumen:25 μl
Specifications
About this item
Nunc™ 384 ShallowWell standard height, black and white plates save costs in samples and reagents by reducing the total well volume to 25 μl. The standard height of the plate allows the use of automation without making adjustments to robotics, readers or liquid handlers.