Reverse osmosis water purification system, PURELAB® Prima
The PURELAB Prima delivers up to 30 litres an hour of type III (RO) water. The system is ideal for feeding ultra-pure water systems and for a wide range of laboratory applications including glassware washing, autoclave feed and for use in environmental cabinets.
- Easy to use; easy to handle, easy to operate and easy to maintain
- All units can be placed on a bench or fixed to the wall using a wall-mounting kit
- Unique membrane configuration: the reverse osmosis elements used are composite spiral polysulfone membranes
- GRID (Graphically Represented Intuitive Display) user interface: simulation of operational mode, treatment flow path and reservoir water level to control the system more effectively
- Safety features: automatic level control, audio-visual alarms, indicator lights to advise consumables need replacing
- Up to 98% rejection of inorganics using reverse osmosis
Feed water: The PRIMA 7, 15 and 20 systems work with a wide range of pressures, ideally between 3 and 6 bar. If water pressure is lower than the minimum required, a boost pump can be installed inside the unit (some units are readily available with an integrated boost pump).
Specifications of water produced:
Minerals: Over 95% removed
Organic materials: Over 99% removed
TOC: Less than 50 ppb
Minerals: Over 95% removed
Organic materials: Over 99% removed
TOC: Less than 0,1 ppm
The PURELAB Prima delivers up to 30 litres an hour of type III (RO) water. The system is ideal for feeding ultra-pure water systems and for a wide range of laboratory applications including glassware washing, autoclave feed and for use in environmental cabinets.
- Easy to use; easy to handle, easy to operate and easy to maintain
- All units can be placed on a bench or fixed to the wall using a wall-mounting kit
- Unique membrane configuration: the reverse osmosis elements used are composite spiral polysulfone membranes
- GRID (Graphically Represented Intuitive Display) user interface: simulation of operational mode, treatment flow path and reservoir water level to control the system more effectively
- Safety features: automatic level control, audio-visual alarms, indicator lights to advise consumables need replacing
- Up to 98% rejection of inorganics using reverse osmosis
Feed water: The PRIMA 7, 15 and 20 systems work with a wide range of pressures, ideally between 3 and 6 bar. If water pressure is lower than the minimum required, a boost pump can be installed inside the unit (some units are readily available with an integrated boost pump).
Specifications of water produced:
Minerals: Over 95% removed
Organic materials: Over 99% removed
TOC: Less than 50 ppb
Minerals: Over 95% removed
Organic materials: Over 99% removed
TOC: Less than 0,1 ppm
- Catalog No:
- 171-0375
- ELGAPR030BPM1
- Modell:
- Prima 30
- Prima 30
- Durchschnittlicher Tagesbedarf:
- 720 l
- 720 l
- Speisewasser:
- Potable mains water supply
- Potable mains water supply
- Produktionsfließrate:
- 30 L/h
- 30 L/h
- TOC-Gehalt:
- <50 ppb
- <50 ppb
- Bakterien:
- <5 CFU/ml
- <5 CFU/ml
- Partikel:
- >99% rejection
- >99% rejection
- B×T×H:
- 410×270×460 mm
- 410×270×460 mm
- Gewicht:
- 13 kg (15 with internal boost pump)
- 13 kg (15 kg with internal boost pump)
Product Family Options
Accessories
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- Bezeichnung
- ELGALC123Carbon filter cartridge, removes free chlorine and particles to protect RO membrane1 * 1 ST - 96,00 €
- Bezeichnung
- Bezeichnung
- Bezeichnung