Charts for temperature, humidity and pressure chart recorders
Catalog # JULS00297
Supplier: JULES RICHARD INSTRUMENTS
Specifications
- Bezeichnung:Diagram sheets for visiotambour range of measurement –15 +40 °C, 7 days
- Seq:04
- VE:100
Specifications
About this item
Recordings 24 h, 7 days or 4×7 days, depending on the model.