Sole-Tester
Brinometer, 0 – 26 %, 290 mm
Artikelnr. 641-1500
Anbieter: Alla France
Springe zu:
Product Details & Documents
Brine is the percentage of saturated solution of salt, usually sodium chloride, in water.
A brinometer is used to measure the percentage of saturated solution of brine.
Dokumente
Spezifikationen
- Bezeichnung:Brinometer
- Länge:290 mm
- Messbereich:0 – 26 %