Caps for cell culture flasks, Nunc™ Cell Factory™
Artikelnr. 734-2152
Lieferant: Thermo Fisher Scientific
Spezifikationen
- Bezeichnung:Zweipositionen-Schraubverschluss für 175/185/500 cm² Flaschen
- VE:100
Spezifikationen
Über diesen Artikel
These caps reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the cell factory system.
Further the development of applications requires a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ accessories.