Aspirator Bottles, with Tube Connector and Drain Cock
Artikelnr. SMIT20382000ML
Lieferant: SMITH SCIENTIFIC
Spezifikationen
- Kapazität:2000 ml
- Seq:02
- NS-Stopfen:19/26
- VE:3
Spezifikationen
Über diesen Artikel
Clear glass with normal ground joint stopper and interchangeable glass drain cock.
- Very good chemical resistance
- Good thermal resistance
- Interchangeable drain cock (replacement cocks can be ordered using Article No. 215-0641)