Centrifugal Filter Units, with Microporous Hydrophilic PTFE Membrane
Supplier: Merck
Certificates
About this item
Ultrafree®-MC centrifugal devices are single-use, disposable PP filters used for removing particulates from aqueous biological solutions. Devices are available with a range of microporous membranes for fast filtration and highly reproducible performance.
- Fast filtration and highly reproducible performance
Applications include clarification of aqueous and some solvent based samples, protein and nucleic acid sample preparation.
Specifications
- Catalog No:
- MILFUFC30LH25
- MILFUFC30LG25
- Filterfläche:
- 0,2 cm²
- 0,2 cm²
- Max. RZB:
- 5000 ×g
- 5000 ×g
- Totvolumen:
- 5 µl
- 5 µl
- Ø×H:
- 10,6×45 mm
- 10,6×45 mm
- Probenvolumen:
- max. 400 µl
- max. 400 µl
- Temperatur:
- 50 °C
- 50 °C