Chemical Duty Pump Maintenance Kit
Catalog # MILFWP61MNT00
Supplier: Merck
Specifications
- Bezeichnung:Chemical duty pump maintenance kit
- VE:1
Specifications
About this item
Delivery: This item includes parts for catalog numbers MILFWP6111560, MILFWP6122050, MILFWP6110060. This item is a kit composed of 1 diaphragm, 1 diaphragm liner, 1 O-ring gasket, 4 head screws, 2 handle screws.