CryoGen® Internal Cap for Tubes, 2D
Internal cap with self standing bottom style, Height: 47,70 mm (±0,34 mm), 2 ml
Artikelnr. BSGMCL2ARBIPS2D
Anbieter: BIOSIGMA
Springe zu:
Product Details & Documents
Human DNA, DNase, RNase, Endotoxins, ATP, PCR inhibitors free certified.
Spezifikationen
- Bezeichnung:Internal cap with self standing bottom style, Height: 47,70 mm (±0,34 mm)
- Größe:2 ml