- Für:PP#5 pipette tip racks
- Kapazität:96 L
- Bezeichnung:CycleBox recycle kit
- B×T×H:400×400×600 mm
- VE:1
Thermo Scientific™ CycleBox recycle kit mail-back program provides a simple and effective solution for recycling Thermo Scientific branded PP#5 non-hazardous pipette tip racks (tips excluded from program).
- Recycle non-hazardous pipette tip racks with CycleBox
- For use with any Thermo Fisher Scientific branded PP#5 pipette tip boxes
- Fill box adhering to list of acceptable materials
- Securely seal box top and bottom
- Request collection by sending an email
- Fill out waste transfer note and arrange a pickup within 3 to 5 working days
- Material is delivered to a facility that converts it back into pellet form
A few simple steps needed to recycle PP#5. Order Cyclebox and fill box with racks only, no tips, adhering to list of acceptable materials. If delivered material is contaminated, a non-conformance procedure may be initiated with potential surcharges, and the recycling box will be returned to you. Once full, seal the box with adhesive tape, close box and securely tape bottom and top. Request a collection by sending an email with Subject line 'ThermoFisher - pipette tip box recycling' to [email protected]. Upon email receipt a response will be provided with following information: Waste Transfer Note, complete Section A and B. Email the signed Waste Transfer Note, along with attached photo of the box and contents to [email protected] to finalize the pickup request. Once information is validated, a pickup arragement will be coordinated within three to five working days. The courier will bring a shipping label and attach it to the box during pickup. The box must be ready to be handed over to the courier on arrival without waiting time. A failed collection will incur a charge for any subsequent attempts. Material is then delivered to a facility and converted back into pellet format so it can be used again.