
Celite® Hyflo® Super Cel, Filterhilfsmittel, Supelco®
Catalog # 1.02688.1000
Supplier: Merck
CAS Number:
Specifications
- Packung:Kunststoffflasche
- VE:1 kg
- Seq:4024
Specifications
Specification Test Results
- Solubility:- 0.001 g/l (20 °C)
- Bulk Density:300 kg/m3
- pH:10 (100 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
Boiling Pt:2200 °C (1013 hPa)
Melting Pt:1710 °C
Density:2.3 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Raumtemperatur
EINECS:272-489-0
:GHS08
CAS Number:68855-54-9
MDL Number:MFCD00132803