TERUFUSION™ Solution Administration Set
2way-spike, 20 drops/ml, 15 µm filter, 2,15 m
Artikelnr. TERUTIPU300WY
Anbieter: TERUMO
Springe zu:
Product Details & Documents
TERUFUSION™ solution administration set (for infusion pump) has unique features meeting the various needs of hospitals.
- Simple
- Efficient use
- PVC, DEHP-free
- AFF clip, Y-site×1, round lock connector
TERUFUSION™ solution administration set for infusion pump is specially designed for use with the TERUFUSION infusion pump for optimal performance and safety.
Spezifikationen
- Bezeichnung:2way-spike, 20 drops/ml, 15 µm filter
- Länge:2,15 m