AuftragsverfolgungKontakt
SURPLUG™ AD Solution Administration Set
SURPLUG™ AD Solution Administration Set
2 way-spike, 20 drops/ml, 0,2 µm filter, 2,5 m
Catalog # TERUSAQTF303NN
Supplier:   TERUMO
SURPLUG™ AD Solution Administration Set
Catalog # TERUSAQTF303NN
Supplier:  TERUMO
SURPLUG™ AD Solution Administration Set
2 way-spike, 20 drops/ml, 0,2 µm filter, 2,5 m
Price
4 250,00 €
Log in for availability
1 * 160 ST
Product Details & Documents
SURPLUG™ AD series infusion system meeting the needs in various departments in a safer and simpler way.
  • A flat top for easier scrubbing
  • Structure for high flow rate
  • Applicable to blood transfusions
  • Round lock connector, PVC-free
  • AFF clip, SURPLUG AD T-Port×3
Using high-quality materials for the system, no need for a dedicated connection adaptor and Terumo's unique structure eliminates drug residues.
Specifications
  • Bezeichnung:
    2 way-spike, 20 drops/ml, 0,2 µm filter
  • Länge:
    2,5 m