TASKI Sani Calc Pur-Eco W3b Washroom Descaler
Artikelnr. DIVY7515243
Lieferant: Diversey
New Product
Spezifikationen
- Kapazität:6 L
- Bezeichnung:Washroom descaler, slightly perfumed
- VE:6 Bottle
Spezifikationen
Über diesen Artikel
Powerful limescale remover for acid resistant hard surfaces.
- Fast acting acidic, based on citric acid formula
- Excellent cleaning performance
- Strong descaling properties
- No scrubbing necessary at high concentration
Certifications: EU Ecolabel (EU Flower).