Buccutite™ Rapid Crosslinked APC Antibody Labelling Kit
Artikelnr. AATB5406
Lieferant: AAT Bioquest
New Product
CAS-Nummer:
Spezifikationen
- VE:2
- Enthält:2 labellings
Spezifikationen
Über diesen Artikel
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ Rapid crosslinked APC antibody labelling kits, designed for large-scale production, offer a convenient and efficient method to label antibodies with APC, PE and iFluor® tandem dyes.
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Lieferumfang: Shipped with blue ice.
Achtung! Bitte beachten: Research use only.