Amplite® Colorimetric Glucose and Sucrose Assay Kit
Catalog # AATB40009
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- VE:100 Tests
- Tests pro Kit:100 tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® Colorimetric Glucose and Sucrose Assay Kit offers a convenient method for measuring glucose and sucrose levels in various biological samples, such as serum, plasma, body fluids, food, tissue extracts, and growth medium.
- Useful for assessing aerobic glycolysis, pyruvate kinase activity, and metabolic profiles in different biological matrices.
- Measured at 570 nm
It is also employed for determining protein and sugar concentrations in food samples, as well as monitoring plasma insulin and glucose levels in murine models.
Delivery: Shipped with blue ice.
Caution: Research use only.