Alconox® Detergens, Pulver
Artikelnr. SIALZ742914-1.8KG
Lieferant: Merck
New Product
Spezifikationen
- Bezeichnung:Alconox® detergent, powder
- Packung:Cartons
- VE:1,8 kg
Spezifikationen
Über diesen Artikel
ALCONOX® is a mixture of a homogeneous blend of sodium linear alkylaryl sulfonate, alcohol sulfate, phosphates, and carbonates. Powdered detergent for general lab cleaning.
- Concentrated and long-lasting
- Biodegradable and readily disposable
- Penetrating wetting power to save
- Alternative for corrosive acids and hazardous solvents
- Easy to rinse, doesn't form interfering residues, and provides reliable results