Supelco® Organischer Referenzstandard, 2-Isopropyläpfelsäure
Supplier: Merck
Certificates
About this item
Specifications
- CAS Nr.:3237-44-3
- Lagerbedingungen:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00075003
- Synonym(s):2-Hydroxy-2-isopropylbernsteinsäure
- Melting point:138...145 °C; 144...146 °C (lit.)
- Formel:HO₂CCH₂C[CH(CH₃)₂](OH)CO₂H
- Molecular weight:176,17