mFluor™ Violet 540-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # AATB25454
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- VE:1 mg
- Bezeichnung:mFluor™ Violet 540-Wheat germ agglutinin (WGA) conjugate
Specifications
About this item
mFluor™ Violet 540 labelled WGA is well-excited by the violet laser, emitting a bright green fluorescence at 535 nm.
- Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.