0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
6-FAM Alkyne
6-FAM Alkyne, FastClick™
6-FAM Alkyne, FastClick™
6-FAM Alkyne, FastClick™
Catalog # AATB72861
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
6-FAM Alkyne, FastClick™
Catalog # AATB72861
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
VE:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
684.75 g/mol
CAS Number:
478801-49-9
Product Details