Anti-NIPA Rabbit Polyclonal Antibody
Catalog # ANTIA94033-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:hNIPA
- Antigen-Symbol:NIPA
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# Q86WB0
- Antigen-Synonyme:Zinc finger C3HC-type protein 1|Nuclear-interacting partner of anaplastic lymphoma kinase|Nuclear interacting partner of anaplastic lymphoma kinase|Zinc finger C3HC type containing 1|Nuclear interacting partner of ALK|Nuclear-interacting partner of ALK|NIPA_HUMAN|zc3hc1
- Lagerungspuffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:55 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transporttemperatur:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human NIPA around the phosphorylation site of Ser354 (amino acids 320-369)
- Reinigung:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Packung:Plastic vial
- VE:100 µG
Specifications
About this item
Rabbit polyclonal antibody to NIPA (phospho Ser354) for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: NIPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat