Anti-ZNF81 Mouse Monoclonal Antibody [clone: PCRP-ZNF81-2G2]
Catalog # ANTIA277657-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:ZNF81
- Antigen-Symbol:ZNF81
- Klonalität:Monoklonal
- Klon:PCRP-ZNF81-2G2
- Konjugation:Unconjugated
- ELISA:Yes
- Durchflusszytometrie:Yes
- Wirt:Mouse
- ImmunFluoreszenz:Yes
- Immunpräzipitation:Yes
- Isotyp:IgG2a
- Reaktivität:Human
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# P51508
- Lagerungspuffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:75,9 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Konzentration:200 µg/ml
- Transporttemperatur:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human ZNF81 protein.
- Reinigung:Protein A/G chromatography
- Packung:Plastic vial
- VE:100 µG
Specifications
About this item
Mouse monoclonal [PCRP-ZNF81-2G2] antibody to ZNF81 for ELISA, WB, IP, Flow Cytometry and IF with samples derived from human.
- Validated Applications: ELISA, WB, IP, Flow Cytometry, IF
- Recommended Dilutions: WB: 1 to 2 µg/ml, IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: ZNF81
Clonality: Monoclonal
Clone: PCRP-ZNF81-2G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human