Anti-CD27 Rabbit Recombinant Antibody [clone: DM58] (Biotin)
Lieferant: ANTIBODIES.COM
New Product
Zertifikate
Über diesen Artikel
Recombinant rabbit monoclonal [DM58] antibody to CD27 (Biotin) for ELISA and Flow Cytometry with samples derived from human.
- Validated Applications: ELISA, Flow Cytometry
- Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100
Type: Primary
Antigen: CD27
Clonality: Recombinant
Clone: DM58
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spezifikationen
- Catalog No:
- ANTIA317407-500
- ANTIA317407-100
- Antigen-Symbol:
- CD27
- CD27
- Antigen-Name:
- CD 27
- CD 27
- Antigen-Synonyme:
- CD27_HUMAN|S152|CD27 antigen|CD27 molecule|CD27L receptor|MGC20393|OTTHUMP00000238557|CD27|LPFS2
- CD27_HUMAN|S152|CD27 antigen|CD27 molecule|CD27L receptor|MGC20393|OTTHUMP00000238557|CD27|LPFS2
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Recombinant
- Recombinant
- Konjugation:
- Biotin
- Biotin
- Klon:
- DM58
- DM58
- Reaktivität:
- Human
- Human
- Wirt:
- Rabbit
- Rabbit
- Gen-ID:
- UniprotID# P26842
- UniprotID# P26842
- Isotyp:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Durchflusszytometrie:
- Yes
- Yes
- Form:
- Lyophilised
- Lyophilised
- Reinigung:
- Affinity chromatography
- Affinity chromatography
- Konzentration:
- Reconstitution dependent
- Reconstitution dependent
- Lagerungspuffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
- Lagertemperatur:
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Transporttemperatur:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Spezifikationen
Frequently Bought Together