Order Entry
Austria
ContactUsLinkComponent
Anti-2B4 Rabbit Recombinant Antibody [clone: DM70]
Anti-2B4 Rabbit Recombinant Antibody [clone: DM70]
Supplier:  ANTIBODIES.COM
New Product
Certificates
undefined
Anti-2B4 Rabbit Recombinant Antibody [clone: DM70]
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Catalog No:
  • ANTIA317394-100
  • ANTIA317394-500
  • Antigen-Symbol:
  • 2B4
  • 2B4
  • Antigen-Name:
  • 2B4
  • 2B4
  • Antikörper-Typ:
  • Primär
  • Primär
  • Klonalität:
  • Recombinant
  • Recombinant
  • Konjugation:
  • Unconjugated
  • Unconjugated
  • Klon:
  • DM70
  • DM70
  • Reaktivität:
  • Human
  • Human
  • Wirt:
  • Rabbit
  • Rabbit
  • Gen-ID:
  • UniprotID# Q9BZW8
  • UniprotID# Q9BZW8
  • Isotyp:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Durchflusszytometrie:
  • Yes
  • Yes
  • Form:
  • Lyophilised
  • Lyophilised
  • Reinigung:
  • Affinity chromatography
  • Affinity chromatography
  • Konzentration:
  • Reconstitution dependent
  • Reconstitution dependent
  • Lagerungspuffer:
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Lagertemperatur:
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Transporttemperatur:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C

Specifications

Frequently Bought Together