Anti-GCLM Rabbit Polyclonal Antibody
Artikelnr. ANTIA95081-100
Lieferant: ANTIBODIES.COM
New Product
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:Gamma ECS regulatory subunit
- Antigen-Symbol:GCLM
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# P48507
- Antigen-Synonyme:GSC light chain|Gamma-glutamylcysteine synthetase regulatory subunit|Glutamate cysteine ligase regulatory subunit|Gamma-ECS regulatory subunit|GLCLR|GCS light chain|Glutamate--cysteine ligase modifier subunit|GCLM|Glutamate--cysteine ligase regulatory subunit
- Lagerungspuffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:30 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transporttemperatur:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human GCSm-gamma (amino acids 42-91)
- Validierte Anwendungen:IHC
- Reinigung:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Packung:Plastic vial
- VE:100 µG
Spezifikationen
Über diesen Artikel
Rabbit polyclonal antibody to GCSm-gamma for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: GCLM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat