Anti-GPCR GPRC5D Rabbit Recombinant Antibody [clone: DM61]
Lieferant: ANTIBODIES.COM
New Product
Zertifikate
Über diesen Artikel
Recombinant rabbit monoclonal [DM61] antibody to GPCR GPRC5D for ELISA, flow cytometry and IHC with samples derived from human.
- Validated applications: ELISA, flow cytometry, IHC
- Recommended dilutions: ELISA: 1:5000 to 10000, flow cytometry: 1:100, IHC: 1:200
Type: Primary
Antigen: GPCR GPRC5D
Clonality: Recombinant
Clone: DM61
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spezifikationen
- Catalog No:
- ANTIA318636-500
- ANTIA318636-100
- Antigen-Symbol:
- GPCR GPRC5D
- GPCR GPRC5D
- Antigen-Name:
- G protein coupled receptor family C group 5 member D
- G protein coupled receptor family C group 5 member D
- Antigen-Synonyme:
- GPC5D_HUMAN|G-protein coupled receptor family C group 5 member D|GPCR GPRC5D
- GPC5D_HUMAN|G-protein coupled receptor family C group 5 member D|GPCR GPRC5D
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Recombinant
- Recombinant
- Konjugation:
- Unconjugated
- Unconjugated
- Klon:
- DM61
- DM61
- Reaktivität:
- Human
- Human
- Wirt:
- Rabbit
- Rabbit
- Gen-ID:
- UniprotID# Q9NZD1
- UniprotID# Q9NZD1
- Isotyp:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemie:
- Yes
- Yes
- Durchflusszytometrie:
- Yes
- Yes
- Form:
- Lyophilised
- Lyophilised
- Reinigung:
- Affinity chromatography
- Affinity chromatography
- Konzentration:
- Reconstitution dependent
- Reconstitution dependent
- Lagerungspuffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Lagertemperatur:
- Shipped at 4 °C. Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Transporttemperatur:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Validierte Anwendungen:
- IHC
- IHC
Spezifikationen
Frequently Bought Together