Order Entry
Austria
ContactUsLinkComponent
Anti-GCLM Rabbit Monoclonal Antibody [clone: ARC0597]
Artikelnr. ANTIA81176-100
Lieferant:  ANTIBODIES.COM
New Product
undefined
Anti-GCLM Rabbit Monoclonal Antibody [clone: ARC0597]
Artikelnr. ANTIA81176-100
Lieferant:  ANTIBODIES.COM
New Product

Spezifikationen

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Gamma ECS regulatory subunit
  • Antigen-Symbol:
    GCLM
  • Klonalität:
    Monoklonal
  • Klon:
    ARC0597
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gen-ID:
    UniprotID# P48507
  • Antigen-Synonyme:
    GSC light chain|Gamma-glutamylcysteine synthetase regulatory subunit|Glutamate cysteine ligase regulatory subunit|Gamma-ECS regulatory subunit|GLCLR|GCS light chain|Glutamate--cysteine ligase modifier subunit|GCLM|Glutamate--cysteine ligase regulatory subunit
  • Lagerungspuffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% Glycerol, 0,05% BSA, and 0,02% Sodium azide
  • Molecular weight:
    30 kDa
  • Sequence:
    LYQWAQVKPNSNQVNLASCCVMPPDLTAFAKQFDIQLLTHNDPKELLSEASFQEALQESIPDIQAHEWVPLWLLRYSVIVKSRGIIKSKGYILQAKRRGS
  • Lagertemperatur:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Transporttemperatur:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 175-274 of human GCLM (P48507)
  • Reinigung:
    Affinity purification
  • Packung:
    Plastic vial
  • VE:
    100 µl

Spezifikationen

Über diesen Artikel

Rabbit monoclonal [ARC0597] antibody to GCLM for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:200

Type: Primary
Antigen: GCLM
Clonality: Monoclonal
Clone: ARC0597
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human