ReadiUse™ Universal Kinase Reaction Stopping Buffer
Catalog # AATB31005
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- VE:100 mL
- Bezeichnung:ReadiUse™ Universal kinase reaction stopping buffer
Specifications
About this item
ReadiUse™ Universal Kinase Reaction Stopping Buffer is an aqueous solution that effectively chelates divalent metal ions.
- Designed for ease of use
- Saves time
This sterile-filtered solution finds wide applicability in biochemistry and molecular biology research.
Caution: For research use only.