Anti-TIMP1 Mouse Recombinant Antibody [clone: rTIMP1/1710]
Catalog # ANTIA250131-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Clgi
- Antigen-Symbol:TIMP1
- Klonalität:Recombinant
- Klon:rTIMP1/1710
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- Isotyp:IgG1 kappa
- Reaktivität:Human
- Form:Liquid
- Gen-ID:UniprotID# P01033
- Antigen-Synonyme:Collagenase inhibitor, Human|Erythroid Potentiating Activity|Fibroblast collagenase inhibitor|EPO|FLJ90373|HCI|Collagenase inhibitor|EPA|Erythroid-potentiating activity
- Lagerungspuffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:23 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Konzentration:200 µg/ml
- Transporttemperatur:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human TIMP1 protein
- Reinigung:Protein A chromatography
- Packung:Plastic vial
- VE:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rTIMP1/1710] antibody to TIMP1 for ELISA with samples derived from Human.
- Validated applications: ELISA
Type: Primary
Antigen: TIMP1
Clonality: Recombinant
Clone: rTIMP1/1710
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human