Anti-GCLM Mouse Monoclonal Antibody [clone: GCLM/4069]
ANTIA316390-100
New Product
- Antikörper-Typ:Primär
- Antigen-Name:Gamma ECS regulatory subunit
- Antigen-Symbol:GCLM
- Klonalität:Monoklonal
- Klon:GCLM/4069
- Konjugation:Unconjugated
- Wirt:Mouse
- ImmunoChemie:Yes
- Isotyp:IgG2a kappa
- Reaktivität:Human
- Form:Liquid
- Gen-ID:UniprotID# P48507
- Antigen-Synonyme:GSC light chain|Gamma-glutamylcysteine synthetase regulatory subunit|Glutamate cysteine ligase regulatory subunit|Gamma-ECS regulatory subunit|GLCLR|GCS light chain|Glutamate--cysteine ligase modifier subunit|GCLM|Glutamate--cysteine ligase regulatory subunit
- Lagerungspuffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:31 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Konzentration:1 mg/ml
- Transporttemperatur:Shipped on blue ice at +4 °C
- Immunogen:Recombinant full-length human GCLM protein.
- Validierte Anwendungen:IHC-P
- Reinigung:Protein A/G chromatography
- Packung:Plastic vial
- VE:100 µG
Mouse monoclonal [GCLM/4069] antibody to GCLM for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: GCLM
Clonality: Monoclonal
Clone: GCLM/4069
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human