Anti-Catalase Rabbit Recombinant Antibody [clone: HPYL/8575R]
Catalog # ANTIA315952-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Cas1
- Antigen-Symbol:Catalase
- Klonalität:Recombinant
- Klon:HPYL/8575R
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG kappa
- Reaktivität:H. pylori
- Form:Liquid
- Antigen-Synonyme:Catalase|MGC138422|EC 1.11.1.6|Cs1|CAT|MGC138424|CATA_HUMAN
- Lagerungspuffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:58 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Konzentration:1 mg/ml
- Transporttemperatur:Shipped on blue ice at +4 °C
- Immunogen:Recombinant Helicobacter pylori Catalase protein fragment (around Amino Acids 323-445). The exact sequence is proprietary.
- Validierte Anwendungen:IHC-P
- Reinigung:Protein A/G chromatography
- Packung:Plastic vial
- VE:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [HPYL/8575R] antibody to Catalase for IHC-P with samples derived from helicobacter pylori.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Catalase
Clonality: Recombinant
Clone: HPYL/8575R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Helicobacter pylori